Anisodamine ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: [(3S,6S)-6-hydroxy-8-methyl-8-azabicyclo[3.2.1]octan-3-yl](2S)-3-hydroxy-2-phenylpropanoate

A276242-100MG A276242-1G A276242-500MG
ALA276242100MGEA 45.51 USD
ALA276242100MG ALA276242-1G ALA276242500MG
Anisodamine ≥97%
Anisodamine
Formula: C₁₇H₂₃NO₄
MW: 305.37 g/mol
Storage Temperature: Freezer
CAS Number: 55869-99-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c220aa82d4e30858f68335bdd0d26cb139173d5&vid=5c220aa82d4e30858f68335bdd0d26cb139173d5&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR