A-1210477 ≥98%
Supplier: ALADDIN SCIENTIFIC CORPORATION
Synonyms:
7-(5-((4-(4-(N,N-dimethylsulphamoyl)piperazin-1-yl)phenoxy)methyl)-1,3-dimethyl-1H-pyrazol-4-yl)-1-(2-morpholinoethyl)-3-(3-(naphthalen-1-yloxy)propyl)-1H-indole-2-carboxylic acid
A276231-10MG
A276231-25MG
A276231-50MG
A276231-5MG
A276231-100MG
ALA276231-10MGEA
112.01
USD
ALA276231-10MG
ALA276231-25MG
ALA276231-50MG
ALA276231-5MG
ALA276231100MG
A-1210477 ≥98%
A-1210477
Shipped at 4°C. Store at -20°C. Store under desiccating conditions.
Formula:
C₄₆H₅₅N₇O₇S MW: 850.04 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD28399118 CAS Number: 1668553-26-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...