(S)-1-(2-(Difluoromethyl)pyridin-4-yl)-4-fluoro-1-(3-(pyrimidin-5-yl)phenyl)-1H-isoindol-3-amine ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: AZD3839

A275977-10MG A275977-1MG A275977-25MG A275977-50MG A275977-5MG
ALA275977-10MGEA 206.06 USD
ALA275977-10MG ALA275977-1MG ALA275977-25MG ALA275977-50MG ALA275977-5MG
(S)-1-(2-(Difluoromethyl)pyridin-4-yl)-4-fluoro-1-(3-(pyrimidin-5-yl)phenyl)-1H-isoindol-3-amine ≥98%, Moligand™
(S)-1-(2-(Difluoromethyl)pyridin-4-yl)-4-fluoro-1-(3-(pyrimidin-5-yl)phenyl)-1H-isoindol-3-amine

Shipped at 4°C. Store at -20°C.

Formula: C₂₄H₁₆F₃N₅
MW: 431.41 g/mol
Storage Temperature: Freezer
MDL Number: MFCD25976574
CAS Number: 1227163-84-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b3dc93d3b435ea9034a37ffdddc77520e1b168b&vid=2b3dc93d3b435ea9034a37ffdddc77520e1b168b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR