6-Amino-9H-purine-9-propanoic acid ≥97%
Supplier: ALADDIN SCIENTIFIC CORPORATION
Synonyms:
3-(6-Amino-9H-purin-9-yl)propionicacid
A275969-100MG
A275969-500MG
ALA275969100MGEA
54.06
USD
ALA275969100MG
ALA275969500MG
6-Amino-9H-purine-9-propanoic acid ≥97%
6-Amino-9H-purine-9-propanoic acid
Formula:
C₈H₉N₅O₂ MW: 207.19 g/mol Storage Temperature: Refrigerator |
CAS Number:
4244-47-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...