ARC 239 dihydrochloride ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2-[2-[4-(2-Methoxyphenyl)-1-piperazinyl]ethyl]-4,4-dimethyl-1,3(2H,4H)-isoquinolinedionedihydrochloride

A275859-100MG A275859-10MG A275859-25MG A275859-50MG A275859-5MG
ALA275859100MGEA 930.91 USD
ALA275859100MG ALA275859-10MG ALA275859-25MG ALA275859-50MG ALA275859-5MG
ARC 239 dihydrochloride ≥98%
ARC 239 dihydrochloride

Store at +4°C. Store under desiccating conditions. The product can be stored for up to 12 months.

Formula: C₂₄H₃₁Cl₂N₃O₃
MW: 480.43 g/mol
Storage Temperature: Freezer
CAS Number: 55974-42-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c756e61f09a6de84f904043a1ab959c878fd09f6&vid=c756e61f09a6de84f904043a1ab959c878fd09f6&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR