N-(4-n-Pentylcinnamoyl)anthranilic acid ≥96%, Moligand™

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: N-(p-Amylcinnamoyl)anthranilic acid, 2-[3-(4-Pentylphenyl)prop-2-enoylamino]benzoicacid, 2-(3-(4-Pentylphenyl)acrylamido)benzoic acid

A275778-100MG A275778-250MG A275778-25MG A275778-50MG
ALA275778100MGEA 121.51 USD
ALA275778100MG ALA275778250MG ALA275778-25MG ALA275778-50MG
N-(4-n-Pentylcinnamoyl)anthranilic acid ≥96%, Moligand™
N-(4-n-Pentylcinnamoyl)anthranilic acid

N-(p-Amylcinnamoyl) anthranilic Acid (ACA), a cell permeable inhibitor of PLA2, has been reported to inhibit α-ketoisocaproic acid and glyceraldehyde-induced insulin secretion, as well as glucose-induced insulin secretion. Additionally, by inhibiting PLA2, ACA consequentially suppresses glucose-induced arachidonic acid formation in cells. Furthermore, research has shown ACA to modulate the activity of different transient receptor potential channels. In particular, ACA's blocking ability of diacylglycerol-activated TRP channels is of special interest since it offers the opportunity to interfere with receptor-induced Ca2+-dependent signaling processes in vascular smooth muscle cells and platelets.

Formula: C₂₁H₂₃NO₃
MW: 337.419 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00211365
CAS Number: 110683-10-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0649e73584c4c2be27ae777768e6ef165e21c2f&vid=f0649e73584c4c2be27ae777768e6ef165e21c2f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR