AMI-1 ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: AMI 1, Sodium 7,7'-(carbonylbis(azanediyl))bis(4-hydroxynaphthalene-2-sulphonate), 7,7'-(Carbonyldiimino)bis[4-hydroxy2-naphthalenesulphonicacid]disodiumsalt, 7,7′-Carbonylbis(azanediyl)bis(4-hydroxynaphthalene-2-sulfonic acid) sodium salt, AMI-1 sodium salt

A275769-25MG A275769-50MG A275769-5MG
ALA275769-25MGEA 150.01 USD
ALA275769-25MG ALA275769-50MG ALA275769-5MG
AMI-1 ≥98%
AMI-1

Store at -20°C. It is important to note that this product is reported to be light sensitive. Store In the Dark. Store under desiccating conditions.

Formula: C₂₁H₁₄N₂Na₂O₉S₂
MW: 548.45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00068249
CAS Number: 20324-87-2 | 134-47-4 (free base)
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6808dcc67e2875139f4b7d20d6cb89340e94e59&vid=b6808dcc67e2875139f4b7d20d6cb89340e94e59&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR