Anandamide (18:2, n-6) ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: Linoleyl ethanolamide, (9Z,12Z)-N-(2-Hydroxyethyl)octadeca-9,12-dienamide, Linoleylethanolamide, N-(2-Hydroxyethyl)linoleamide, n-linoleoylethanolamine, Linoleamide MEA, Linoleic ethanolamide

A275712-100MG A275712-25MG A275712-5MG
ALA275712100MGEA 1097.16 USD
ALA275712100MG ALA275712-25MG ALA275712-5MG
Anandamide (18:2, n-6) ≥98%
Anandamide (18:2, n-6)

Store at -20°C. Store under desiccating conditions. The product can be stored for up to 12 months.

Formula: C₂₀H₃₇NO₂
MW: 323.5 g/mol
Storage Temperature: Freezer
CAS Number: 68171-52-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=54f79fe74be819f32b4857ed79391026f0d97f77&vid=54f79fe74be819f32b4857ed79391026f0d97f77&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR