APS-2-79 hydrochloride ≥98%

Supplier: ALADDIN SCIENTIFIC

Synonyms: APS-2-79, 6,7-dimethoxy-N-(2-methyl-4-phenoxyphenyl)quinazolin-4-aminehydrochloride, 6,7-Dimethoxy-N-(2-methyl-4-phenoxyphenyl)quinazolin-4-amine hydrochloride, 6,7-dimethoxy-N-(2-methyl-4-phenoxyphenyl)quinazolin-4-amine hydrochloride

A275508-50MG A275508-5MG A275508-10MG A275508-25MG
ALA275508-50MGEA 637.36 USD
ALA275508-50MG ALA275508-5MG ALA275508-10MG ALA275508-25MG
APS-2-79 hydrochloride ≥98%
APS-2-79 hydrochloride

Shipped at 4°C. Store at -20°C. Store In the Dark. Store under desiccating conditions.

Formula: C₂₃H₂₂ClN₃O₃
MW: 423.89 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30536203
CAS Number: 2002381-31-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ebe40c6ff88258e46fd55640c0873cc1c43e991&vid=5ebe40c6ff88258e46fd55640c0873cc1c43e991&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR