A-1331852 ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: 3-(1-(adamantan-1-ylmethyl)-5-methyl-1H-pyrazol-4-yl)-6-(8-(benzo[d]thiazol-2-ylcarbamoyl)-3,4-dihydroisoquinolin-2(1H)-yl) picolinic acid, 3-(1-(Adamantan-1-ylmethyl)-5-methyl-1H-pyrazol-4-yl)-6-(8-(benzo[d]thiazol-2-ylcarbamoyl)-3,4-dihydroisoquinolin-2(1H)-yl)picolinic acid

A275493-10MG A275493-25MG A275493-50MG A275493-5MG
ALA275493-10MGEA 213.66 USD
ALA275493-10MG ALA275493-25MG ALA275493-50MG ALA275493-5MG
A-1331852 ≥98%, Moligand™
A-1331852

Shipped at 4°C. Store at -20°C. Store under desiccating conditions.

Formula: C₃₈H₃₈N₆O₃S
MW: 658.81 g/mol
Storage Temperature: Freezer
MDL Number: MFCD30532662
CAS Number: 1430844-80-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79f41a684bad9cac58bb77643e3ad47f30339587&vid=79f41a684bad9cac58bb77643e3ad47f30339587&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR