Acetyl-L-Carnitine ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: (3R)-3-acetyloxy-4-(trimethylazaniumyl)butanoate

A275415-1G A275415-5G
ALA275415-1GEA 45.51 USD
ALA275415-1G ALA275415-5G
Acetyl-L-Carnitine ≥98%, Moligand™
Acetyl-L-Carnitine

Store at Room Temperature. The product can be stored for up to 12 months.

Formula: C₉H₁₇NO₄
MW: 203.24 g/mol
Storage Temperature: Ambient
CAS Number: 3040-38-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a11f3f3f3a30a4c1aec49264f3659a4c8005c54&vid=2a11f3f3f3a30a4c1aec49264f3659a4c8005c54&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR