AH-6809 ≥97%, Moligand™

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: AH 6809, 6-Isopropoxy-9-oxo-9H-xanthene-2-carboxylic acid

A275394-100MG A275394-10MG A275394-50MG
ALA275394100MGEA 806.46 USD
ALA275394100MG ALA275394-10MG ALA275394-50MG
AH-6809 ≥97%, Moligand™
AH-6809

Application:AH6809 has been used:as a prostaglandin E2 receptor 1 & 2 (EP1/2) antagonist to analyze its effects on cyclooxygenase-2 /prostaglandin E2 (COX-2/PGE2) signaling in the angiogenic feedback of endothelial cells to hypoxiaas an EP2 antagonist to study its effects on tumor angiogenesis in human prostate cancer cell linesas an EP2 antagonist to analyze its effects on overexpression of amyloid precursor protein (APP)

Formula: C₁₇H₁₄O₅
MW: 298.3 g/mol
Storage Temperature: Freezer
MDL Number: MFCD08056083
CAS Number: 33458-93-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4da114d921caea99413e5074d4d31907a93719ae&vid=4da114d921caea99413e5074d4d31907a93719ae&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR