5-(4-Hydroxyphenethyl)benzene-1,3-diol ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: Dihydroresveratrol, 3,4′,5-Trihydroxybibenzyl, 3,5,4′-Trihydroxybibenzyl, 5-[2-(4-Hydroxyphenyl)ethyl]benzene-1,3-diol, Dihydroresveratrol, alpha,beta-Dihydroresveratrol

A275297-100MG A275297-10MG A275297-1G A275297-250MG A275297-25MG
ALA275297100MGEA 1292.86 USD
ALA275297100MG ALA275297-10MG ALA275297-1G ALA275297250MG ALA275297-25MG
5-(4-Hydroxyphenethyl)benzene-1,3-diol ≥98%
5-(4-Hydroxyphenethyl)benzene-1,3-diol
Formula: C₁₄H₁₄O₃
MW: 230.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25973128
CAS Number: 58436-28-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99c0a9e19b8588912abb7faf2e93cac8bcc349c7&vid=99c0a9e19b8588912abb7faf2e93cac8bcc349c7&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR