AUNP-12 TFA ≥98%

Supplier: Aladdin Scientific

A275285-10MG A275285-25MG A275285-2MG A275285-5MG
ALA275285-10MGEA 788.41 USD
ALA275285-10MG ALA275285-25MG ALA275285-2MG ALA275285-5MG
AUNP-12 TFA ≥98%
AUNP-12 TFA
Formula: C₁₄₂H₂₂₆N₄₀O₄₈ (free base)
Storage Temperature: Freezer
CAS Number: 1353563-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR