ASB-14 ≥97%
Supplier: ALADDIN SCIENTIFIC
Synonyms:
Amidosulphobetaine-14
A275232-250MG
A275232-1G
ALA275232250MGEA
37.91
USD
ALA275232250MG
ALA275232-1G
ASB-14 ≥97%
ASB-14
Store at +4°C. The product can be stored for up to 12 months.
Formula:
C₂₂H₄₆N₂O₄S MW: 434.68 g/mol Storage Temperature: Refrigerator |
CAS Number:
216667-08-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...