AdipoRon ≥98%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
Synonyms:
SC-396658, 2-(4-Benzoylphenoxy)-N-[1-(phenylmethyl)-4-piperidinyl]acetamide, 2-(4-Benzoylphenoxy)-N-(1-benzylpiperidin-4-yl)acetamide
A275223-100MG
A275223-10MG
A275223-250MG
A275223-50MG
ALA275223100MGEA
390.36
USD
ALA275223100MG
ALA275223-10MG
ALA275223250MG
ALA275223-50MG
AdipoRon ≥98%
AdipoRon
Store at -20°C. Store under desiccating conditions. The product can be stored for up to 12 months.
Formula:
C₂₇H₂₈N₂O₃ MW: 428.52 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD08271060 CAS Number: 924416-43-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...