2-Acetamido-2,4-dideoxy-4-fluoro-alpha-D-glucopyranose ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: 2-Acetamido-2,4-dideoxy-4-fluoro-α-D-glucopyranose

A275122-50MG A275122-5MG
ALA275122-50MGEA 1254.86 USD
ALA275122-50MG ALA275122-5MG
2-Acetamido-2,4-dideoxy-4-fluoro-alpha-D-glucopyranose ≥97%
2-Acetamido-2,4-dideoxy-4-fluoro-alpha-D-glucopyranose
Formula: C₈H₁₄FNO₅
MW: 223.2 g/mol
Storage Temperature: Freezer
CAS Number: 129728-87-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c73096fd3ed2cdb0b605111eeb69fc43e59739e&vid=1c73096fd3ed2cdb0b605111eeb69fc43e59739e&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR