1-(4-(4-Amino-1-isopropyl-1H-pyrazolo[3,4-d]pyrimidin-3-yl)phenyl)-3-(2-fluoro-5-(trifluoromethyl)phenyl)urea ≥99%

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: AD80

A275119-100MG A275119-10MG A275119-25MG A275119-2MG A275119-50MG A275119-5MG
ALA275119100MGEA 2171.61 USD
ALA275119100MG ALA275119-10MG ALA275119-25MG ALA275119-2MG ALA275119-50MG ALA275119-5MG
1-(4-(4-Amino-1-isopropyl-1H-pyrazolo[3,4-d]pyrimidin-3-yl)phenyl)-3-(2-fluoro-5-(trifluoromethyl)phenyl)urea ≥99%
1-(4-(4-Amino-1-isopropyl-1H-pyrazolo[3,4-d]pyrimidin-3-yl)phenyl)-3-(2-fluoro-5-(trifluoromethyl)phenyl)urea

AD80, a multi kinase inhibitor, has strong activity on human RET (c-RET), BRAF, S6K and SRC, but weak activity on mTOR than ad57 or ad58. Its IC50 value for RET (c-RET) is 4 nm.

Formula: C₂₂H₁₉F₄N₇O
MW: 473.43 g/mol
Storage Temperature: Freezer
MDL Number: MFCD31382193
CAS Number: 1384071-99-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dbd02acd050b188988f9129bcb7622d3e7b5f5a2&vid=dbd02acd050b188988f9129bcb7622d3e7b5f5a2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR