Amifostine ≥98%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
A274806-100MG
A274806-10MG
A274806-250MG
A274806-50MG
ALA274806100MGEA
460.66
USD
ALA274806100MG
ALA274806-10MG
ALA274806250MG
ALA274806-50MG
Amifostine ≥98%
Amifostine
Store at +4°C. Store under desiccating conditions. The product can be stored for up to 12 months.
Formula:
C₅H₁₅N₂O₃PS MW: 214.22 g/mol Storage Temperature: Freezer |
CAS Number:
20537-88-6 |
|
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...