Tyrphostin AG-126 ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 2-[(3-Hydroxy-4-nitrophenyl)methylidene]propanedinitrile, Tyrphostin AG 126, 2-(3-Hydroxy-4-nitrobenzylidene)malononitrile, AG-126, (3-Hydroxy-4-nitrobenzylidene)malononitrile

A274763-100MG A274763-10MG A274763-25MG A274763-5MG
ALA274763100MGEA 947.06 USD
ALA274763100MG ALA274763-10MG ALA274763-25MG ALA274763-5MG
Tyrphostin AG-126 ≥98%
Tyrphostin AG-126

Store at Room Temperature. The product can be stored for up to 12 months.

Formula: C₁₀H₅N₃O₃
MW: 215.168 g/mol
Melting Pt: 184 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00236448
CAS Number: 118409-62-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4acc941079f9a8892afb94683ef1f3bbcd02b0da&vid=4acc941079f9a8892afb94683ef1f3bbcd02b0da&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR