N1,N2-Dibenzhydrylethane-1,2-diamine dihydrochloride ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: AMN 082 dihydrochloride, N,N′-Dibenzhydrylethane-1,2-diamine dihydrochloride, AMN082 dihydrochloride, N,N'-Dibenzhydrylethane-1,2-diaminedihydrochloride, AMN082

A274646-50MG A274646-5MG A274646-100MG A274646-10MG
ALA274646-50MGEA 873.91 USD
ALA274646-50MG ALA274646-5MG ALA274646100MG ALA274646-10MG
N1,N2-Dibenzhydrylethane-1,2-diamine dihydrochloride ≥98%, Moligand™
N1,N2-Dibenzhydrylethane-1,2-diamine dihydrochloride

AMN082 dihydrochloride, a selective,  and brain penetrant mGluR7 agonist, directly activates receptor signaling via an allosteric site in the transmembrane domain. AMN082 potently inhibits cAMP accumulation and stimulates GTPγS binding (EC50 values, 64-290 nM) at transfected mammalian cells expressing mGluR7. AMN082 shows selectivity over other mGluR subtypes and selected ionotropic glutamate receptors.

Formula: C₂₈H₃₀Cl₂N₂
MW: 465.4572 g/mol
Storage Temperature: Freezer
MDL Number: MFCD08702723
CAS Number: 97075-46-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62aa9e497bd9d1e9ecf01469eadec3894c0223d8&vid=62aa9e497bd9d1e9ecf01469eadec3894c0223d8&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR