Alsterpaullone ≥98%, Moligand™
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
7,12-Dihydro-9-nitroindolo[3,2-d][1]benzazepin-6(5H)-one
A274619-100MG
A274619-25MG
A274619-50MG
A274619-5MG
ALA274619100MGEA
2190.61
USD
ALA274619100MG
ALA274619-25MG
ALA274619-50MG
ALA274619-5MG
Alsterpaullone ≥98%, Moligand™
Alsterpaullone
Formula:
C₁₆H₁₁N₃O₃ Storage Temperature: Freezer |
MDL Number:
MFCD02683579 CAS Number: 237430-03-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...