8-Amino-2-naphthol ≥94%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 1-Amino-7-naphthol

A196891-1G A196891-25G A196891-5G
ALA196891-1GEA 19.86 USD
ALA196891-1G ALA196891-25G ALA196891-5G
8-Amino-2-naphthol ≥94%
8-Amino-2-naphthol
Formula: C₁₀H₉NO
MW: 159.19 g/mol
Melting Pt: 206-210°C
Density: 1.281 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00004031
CAS Number: 118-46-7
EINECS: 204-252-4
UN: 2811
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0fbf58c3a211733a04a1d1ecac55fe1656e503f8&vid=0fbf58c3a211733a04a1d1ecac55fe1656e503f8&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR