4-Aminobenzenesulfonic acid monosodium salt ≥98%

Supplier: ALADDIN SCIENTIFIC

Synonyms: Sodium p-Anilinesulphonate Hydrate, 4-sulphoaniline Sodium Salt Hydrate, sulphanilic acid Sodium Salt Hydrate, Sodium 4-Aminobenzenesulphonate Hydrate, 4-Amino-benzenesulphonic acid monosodium salt

A196888-100G A196888-25G
ALA196888-100GEA 226.01 USD
ALA196888-100G ALA196888-25G
4-Aminobenzenesulfonic acid monosodium salt ≥98%
4-Aminobenzenesulfonic acid monosodium salt
Formula: C₆H₆NNaO₃S
MW: 195.168 g/mol
Storage Temperature: Ambient
CAS Number: 515-74-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9de36e47e4b60b4289aa37b491132713216f629&vid=b9de36e47e4b60b4289aa37b491132713216f629&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR