4-Amino-2,6-dibromophenol 97

Supplier: ALADDIN SCIENTIFIC
Warning

A196361-50G A196361-5G A196361-1G A196361-25G
ALA196361-50GEA 849.21 USD
ALA196361-50G ALA196361-5G ALA196361-1G ALA196361-25G
4-Amino-2,6-dibromophenol 97
4-Amino-2,6-dibromophenol
Formula: C₆H₅Br₂NO
MW: 266.92
Boiling Pt: 324
Melting Pt: 192
MDL Number: MFCD00007874
CAS Number: 609-21-2
EINECS: 210-185-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=031f93069efab24168e8290ac563ffb7b939bd9d&vid=031f93069efab24168e8290ac563ffb7b939bd9d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR