Antimony(III) iodide ≥99.99% (metals basis), PrimorTrace™

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: Antimony triiodide

A196220-1G A196220-200MG A196220-5G
ALA196220-1GEA 36.01 USD
ALA196220-1G ALA196220200MG ALA196220-5G
Antimony(III) iodide ≥99.99% (metals basis), PrimorTrace™
Antimony(III) iodide

Antimony(III) iodide is used as a dopant in the preparation of thermoelectric materials.

Formula: I₃Sb
MW: 502.47 g/mol
Boiling Pt: 420°C
Melting Pt: 168°C
Density: 4.92 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00016322
CAS Number: 7790-44-5
EINECS: 232-205-8
UN: 3260
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80063a3af442b17e059a0980fb72bd6265ba1640&vid=80063a3af442b17e059a0980fb72bd6265ba1640&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR