4-Acetyl-N-methylaniline ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 1-(4-(Methylamino)phenyl)ethanone, 4'-(Methylamino)acetophenone

A196189-100MG A196189-1G A196189-250MG A196189-25G A196189-5G
ALA196189100MGEA 10.36 USD
ALA196189100MG ALA196189-1G ALA196189250MG ALA196189-25G ALA196189-5G
4-Acetyl-N-methylaniline ≥97%
4-Acetyl-N-methylaniline
Formula: C₉H₁₁NO
MW: 149.193 g/mol
Melting Pt: 105°C
Storage Temperature: Ambient
MDL Number: MFCD09261160
CAS Number: 17687-47-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0671b5837671eb40720771049f3c2630812e05d&vid=b0671b5837671eb40720771049f3c2630812e05d&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR