4-Amino-5-methylpyridin-2-ol ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

A196047-1G A196047-250MG A196047-25G A196047-5G
ALA196047-1GEA 88.26 USD
ALA196047-1G ALA196047250MG ALA196047-25G ALA196047-5G
4-Amino-5-methylpyridin-2-ol ≥98%
4-Amino-5-methylpyridin-2-ol
Formula: C₆H₈N₂O
MW: 124.14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12546373
CAS Number: 95306-64-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b1bacc9e4a5b59405092482135b685e10a8fe41&vid=0b1bacc9e4a5b59405092482135b685e10a8fe41&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR