2-Aminothiazole-5-carbaldehyde hydrochloride ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A195849-1G A195849-250MG
ALA195849-1GEA 30.31 USD
ALA195849-1G ALA195849250MG
2-Aminothiazole-5-carbaldehyde hydrochloride ≥95%
2-Aminothiazole-5-carbaldehyde hydrochloride
Formula: C₄H₅ClN₂OS
Storage Temperature: Ambient
MDL Number: MFCD09991571
CAS Number: 920313-27-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd27f7171b63a1d0529005da5bddf6d4dd90e899&vid=bd27f7171b63a1d0529005da5bddf6d4dd90e899&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR