3-Amino-5-chlorophenol ≥97%
Supplier: ALADDIN SCIENTIFIC
A195521-1G
A195521-250MG
A195521-5G
ALA195521-1GEA
139.56
USD
ALA195521-1G
ALA195521250MG
ALA195521-5G
3-Amino-5-chlorophenol ≥97%
3-Amino-5-chlorophenol
Formula:
C₆H₆ClNO |
MDL Number:
MFCD16999375 CAS Number: 883195-40-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...