2-Amino-5-bromo-3-methoxybenzoic acid ≥95%
Supplier: Aladdin Scientific
|
Warning
|
A195399-1G
A195399-250MG
A195399-5G
ALA195399-1GEA
47.41
USD
ALA195399-1G
ALA195399250MG
ALA195399-5G
2-Amino-5-bromo-3-methoxybenzoic acid ≥95%
2-Amino-5-bromo-3-methoxybenzoic acid
Formula:
C₈H₈BrNO₃ Storage Temperature: Ambient |
MDL Number:
MFCD11101005 CAS Number: 864293-44-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...