2-(7-Amino-2-oxo-2H-chromen-4-yl)acetic acid ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A195313-1G A195313-250MG A195313-5G
ALA195313-1GEA 141.46 USD
ALA195313-1G ALA195313250MG ALA195313-5G
2-(7-Amino-2-oxo-2H-chromen-4-yl)acetic acid ≥95%
2-(7-Amino-2-oxo-2H-chromen-4-yl)acetic acid
Formula: C₁₁H₉NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD13185880
CAS Number: 85157-21-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a794d018c82cc6dfb9bc5f0930d4e185efa419e8&vid=a794d018c82cc6dfb9bc5f0930d4e185efa419e8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR