2-((3-Amino-4-methoxyphenyl)amino)ethanol ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A195234-100MG A195234-1G A195234-250MG A195234-25MG
ALA195234100MGEA 47.41 USD
ALA195234100MG ALA195234-1G ALA195234250MG ALA195234-25MG
2-((3-Amino-4-methoxyphenyl)amino)ethanol ≥98%
2-((3-Amino-4-methoxyphenyl)amino)ethanol
Formula: C₉H₁₄N₂O₂
Boiling Pt: 401.8 °C
Melting Pt: 76-77 °C
Density: 1.221 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD10566418
CAS Number: 83763-47-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c2cd53c6a8e1f8b9fbcdfe71e8fdd51e7fd13f9&vid=5c2cd53c6a8e1f8b9fbcdfe71e8fdd51e7fd13f9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR