4-Amino-N-phenylbenzamide ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A195050-1G A195050-250MG A195050-5G
ALA195050-1GEA 39.81 USD
ALA195050-1G ALA195050250MG ALA195050-5G
4-Amino-N-phenylbenzamide ≥97%
4-Amino-N-phenylbenzamide
Formula: C₁₃H₁₂N₂O
Melting Pt: 127-131 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00017107
CAS Number: 782-45-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7de09aad9061092f9993c1aacb7b3640b64658dd&vid=7de09aad9061092f9993c1aacb7b3640b64658dd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR