1-(4-Aminophenyl)-4-(4-hydroxyphenyl)piperazine 98
Supplier: ALADDIN SCIENTIFIC
A194882-10G
A194882-25G
A194882-5G
ALA194882-10GEA
14.16
USD
ALA194882-10G
ALA194882-25G
ALA194882-5G
1-(4-Aminophenyl)-4-(4-hydroxyphenyl)piperazine 98
1-(4-Aminophenyl)-4-(4-hydroxyphenyl)piperazine
Formula:
C₁₆H₁₉N₃O MW: 269.34 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD02093082 CAS Number: 74853-08-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...