1-(6-Aminohexyl)-1H-pyrrole-2,5-dione 2,2,2-trifluoroacetate ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A194795-100MG A194795-1G A194795-250MG A194795-25MG A194795-5G
ALA194795100MGEA 70.21 USD
ALA194795100MG ALA194795-1G ALA194795250MG ALA194795-25MG ALA194795-5G
1-(6-Aminohexyl)-1H-pyrrole-2,5-dione 2,2,2-trifluoroacetate ≥97%
1-(6-Aminohexyl)-1H-pyrrole-2,5-dione 2,2,2-trifluoroacetate
Formula: C₁₀H₁₆N₂O₂·CF₃CO₂H
Storage Temperature: Refrigerator
MDL Number: MFCD11519191
CAS Number: 731862-92-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84646d533bcea94204a2c1d4422df133e18e38f9&vid=84646d533bcea94204a2c1d4422df133e18e38f9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR