3'-Amino-2'-hydroxyacetophenone ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A194714-5G A194714-100G A194714-25G
ALA194714-5GEA 9.41 USD
ALA194714-5G ALA194714-100G ALA194714-25G
3'-Amino-2'-hydroxyacetophenone ≥98%
3'-Amino-2'-hydroxyacetophenone
Formula: C₈H₉NO₂
MDL Number: MFCD03428566
CAS Number: 70977-72-9
EINECS: 676-239-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4397b443c124adc4d758ce82019fa426bcb9be56&vid=4397b443c124adc4d758ce82019fa426bcb9be56&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR