Aluminum monostearate

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: Dihydroxy(stearoyloxy)aluminum, Aluminum monostearate technical(Hard and soft aluminium mixture), Aluminum dihydroxide stearate

A194692-100G A194692-25G
ALA194692-100GEA 59.76 USD
ALA194692-100G ALA194692-25G
Aluminum monostearate
Aluminum monostearate
Formula: C₁₈H₃₇AlO₄
MW: 344.4655 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00019932
CAS Number: 7047-84-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7bdb3cc3802917f4936938ac1ae04c2a387571c8&vid=7bdb3cc3802917f4936938ac1ae04c2a387571c8&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR