DL-β-(3-Chlorophenyl)alaninol ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 3-Amino-3-(3-chlorophenyl)propan-1-ol, 3-Amino-3-(3-chlorophenyl)-propan-1-ol

A194578-1G A194578-250MG
ALA194578-1GEA 448.31 USD
ALA194578-1G ALA194578250MG
DL-β-(3-Chlorophenyl)alaninol ≥97%
DL-β-(3-Chlorophenyl)alaninol
Formula: C₉H₁₂ClNO
Storage Temperature: Ambient
MDL Number: MFCD03412790
CAS Number: 68208-25-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b11fa6efd9909d8eda210790e0ad340154964eed&vid=b11fa6efd9909d8eda210790e0ad340154964eed&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR