2-(Azetidin-1-yl)ethanol ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION

A194559-1G A194559-250MG A194559-50MG
ALA194559-1GEA 1142.76 USD
ALA194559-1G ALA194559250MG ALA194559-50MG
2-(Azetidin-1-yl)ethanol ≥98%
2-(Azetidin-1-yl)ethanol
Formula: C₅H₁₁NO
MW: 101.147 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09743797
CAS Number: 67896-18-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb8308165527137f1356e3c36df354fe4d6e7603&vid=cb8308165527137f1356e3c36df354fe4d6e7603&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR