1-(5-Amino-2-fluorophenyl)ethanone ≥98%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
Synonyms:
5'-Amino-2'-fluoroacetophenone
A194531-5G
A194531-100MG
A194531-1G
A194531-250MG
ALA194531-5GEA
1458.16
USD
ALA194531-5G
ALA194531100MG
ALA194531-1G
ALA194531250MG
1-(5-Amino-2-fluorophenyl)ethanone ≥98%
1-(5-Amino-2-fluorophenyl)ethanone
Formula:
C₈H₈FNO MW: 153.16 g/mol |
MDL Number:
MFCD02090052 CAS Number: 67500-19-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...