5-Amino-6-methyl-1,3-dihydro-benzoimidazol-2-one ≥95%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
5-Amino-6-methyl-1H-benzo[d]imidazol-2(3H)-one, 5-Amino-6-methylbenzimidazolone
A194501-1G
A194501-250MG
ALA194501-1GEA
46.46
USD
ALA194501-1G
ALA194501250MG
5-Amino-6-methyl-1,3-dihydro-benzoimidazol-2-one ≥95%
5-Amino-6-methyl-1,3-dihydro-benzoimidazol-2-one
Formula:
C₈H₉N₃O |
MDL Number:
MFCD02684428 CAS Number: 67014-36-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...