2-Amino-3-chlorobenzylalkohol ≥98%

Supplier: ALADDIN SCIENTIFIC

A194203-1G A194203-250MG A194203-5G
ALA194203-1GEA 89.21 USD
ALA194203-1G ALA194203250MG ALA194203-5G
2-Amino-3-chlorobenzylalkohol ≥98%
2-Amino-3-chlorobenzylalkohol
Formula: C₇H₈ClNO
Storage Temperature: Refrigerator
MDL Number: MFCD13193432
CAS Number: 61487-25-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e9938e6eb4741ff0501fd1314dcec666b76839c&vid=4e9938e6eb4741ff0501fd1314dcec666b76839c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR