5-Amino-3-(trifluoromethyl)picolinonitrile ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A194013-1G A194013-5G
ALA194013-1GEA 17.96 USD
ALA194013-1G ALA194013-5G
5-Amino-3-(trifluoromethyl)picolinonitrile ≥98%
5-Amino-3-(trifluoromethyl)picolinonitrile
Formula: C₇H₄F₃N₃
MW: 187.122 g/mol
MDL Number: MFCD17010171
CAS Number: 573762-62-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1b0cdbee0278b5fe094e34ab2026b9409efc312&vid=d1b0cdbee0278b5fe094e34ab2026b9409efc312&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR