4-Aminobenzenesulfonic acid monosodium salt ≥97%
Supplier: ALADDIN SCIENTIFIC CORPORATION
Synonyms:
Sodium p-Anilinesulphonate Hydrate, 4-sulphoaniline Sodium Salt Hydrate, sulphanilic acid Sodium Salt Hydrate, Sodium 4-Aminobenzenesulphonate Hydrate, 4-Amino-benzenesulphonic acid monosodium salt
A193694-100G
A193694-25G
ALA193694-100GEA
49.31
USD
ALA193694-100G
ALA193694-25G
4-Aminobenzenesulfonic acid monosodium salt ≥97%
4-Aminobenzenesulphonic acid monosodium salt
Formula:
C₆H₆NNaO₃S MW: 195.168 g/mol Storage Temperature: Ambient |
CAS Number:
515-74-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...