Azulen-2-amine ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A193621-100MG A193621-1G A193621-25MG A193621-500MG
ALA193621100MGEA 112.01 USD
ALA193621100MG ALA193621-1G ALA193621-25MG ALA193621500MG
Azulen-2-amine ≥97%
Azulen-2-amine
Formula: C₁₀H₉N
Storage Temperature: Refrigerator
MDL Number: MFCD18072913
CAS Number: 50472-20-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62868dd8a20b1dc8ec9edf0ca4709baae8445a9f&vid=62868dd8a20b1dc8ec9edf0ca4709baae8445a9f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR