2-Amino-3-Hydroxyphenazine ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 3-amino-phenazin-2-ol, Hydroxy-2-amino-3-phenazin, 3-Amino-2-phenazinol

A193486-1G A193486-250MG A193486-5G
ALA193486-1GEA 75.91 USD
ALA193486-1G ALA193486250MG ALA193486-5G
2-Amino-3-Hydroxyphenazine ≥98%
2-Amino-3-Hydroxyphenazine
Formula: C₁₂H₉N₃O
Storage Temperature: Ambient
CAS Number: 4569-77-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3dac8948361a1a3d180276dc5bd0a8db0a409eb7&vid=3dac8948361a1a3d180276dc5bd0a8db0a409eb7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR