4-Amino-3,5-dibromobenzoic acid ≥98%
Supplier: Aladdin Scientific
A193319-1G
A193319-25G
A193319-5G
ALA193319-1GEA
19.86
USD
ALA193319-1G
ALA193319-25G
ALA193319-5G
4-Amino-3,5-dibromobenzoic acid ≥98%
4-Amino-3,5-dibromobenzoic acid
Formula:
C₇H₅Br₂NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00075845 CAS Number: 4123-72-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...