(2-Amino-6-chlorophenyl)methanol ≥97%

Supplier: ALADDIN SCIENTIFIC

A193209-1G A193209-250MG A193209-25G A193209-5G
ALA193209-1GEA 14.16 USD
ALA193209-1G ALA193209250MG ALA193209-25G ALA193209-5G
(2-Amino-6-chlorophenyl)methanol ≥97%
(2-Amino-6-chlorophenyl)methanol
Formula: C₇H₈ClNO
MDL Number: MFCD09029932
CAS Number: 39885-08-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06fd6fd4615929183b34bc8704d881f0637b2ff6&vid=06fd6fd4615929183b34bc8704d881f0637b2ff6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR