2-Aminoisophthalic acid 95

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Aminoisophthalic Acid

A193185-100G A193185-25G A193185-5G
ALA193185-100GEA 331.46 USD
ALA193185-100G ALA193185-25G ALA193185-5G
2-Aminoisophthalic acid 95
2-Aminoisophthalic acid
Formula: C₈H₇NO₄
MW: 181.15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00817808
CAS Number: 39622-79-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ab52a237080d175d01da8e55ab87bac98f8e6bd&vid=0ab52a237080d175d01da8e55ab87bac98f8e6bd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR